Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600024901.mm.1 |
TC0600003196.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600024901.mm.1 |
NM_001080979.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024901.mm.1 |
NM_001080979.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024901.mm.1 |
NM_011567.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024901.mm.1 |
NM_011567.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024903.mm.1 |
TC0600003196.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600024903.mm.1 |
NM_001080979.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024903.mm.1 |
NM_001080979.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024903.mm.1 |
NM_011567.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024903.mm.1 |
NM_011567.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024905.mm.1 |
TC0600003196.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600024905.mm.1 |
NM_001080979.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024905.mm.1 |
NM_001080979.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024905.mm.1 |
NM_011567.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024905.mm.1 |
NM_011567.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024907.mm.1 |
TC0600003196.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600024907.mm.1 |
NM_001080979.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024907.mm.1 |
NM_001080979.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024907.mm.1 |
NM_011567.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024907.mm.1 |
NM_011567.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024899.mm.1 |
TC0600003196.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600024899.mm.1 |
NM_001080979.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024899.mm.1 |
NM_001080979.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024899.mm.1 |
NM_011567.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024899.mm.1 |
NM_011567.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024900.mm.1 |
TC0600003196.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600024900.mm.1 |
NM_001080979.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024900.mm.1 |
NM_001080979.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024900.mm.1 |
NM_011567.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024900.mm.1 |
NM_011567.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024906.mm.1 |
TC0600003196.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600024906.mm.1 |
NM_001080979.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024906.mm.1 |
NM_001080979.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024906.mm.1 |
NM_011567.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024906.mm.1 |
NM_011567.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024904.mm.1 |
TC0600003196.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600024904.mm.1 |
NM_001080979.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024904.mm.1 |
NM_001080979.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024904.mm.1 |
NM_011567.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024904.mm.1 |
NM_011567.2 |
Refseq |