Assay Details
| Gene Symbol: Cracr2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033464.3 | NP_001028636.1 | 3 - 4 | 472 | 95 | |
| GenBank mRNA | AK143832.1 | - | 3 - 4 | 473 | 95 | |
| BC150869.1 | - | 3 - 4 | 550 | 9087780 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cracr2a
Genomic Map