Assay Details
Gene Symbol: Sec13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024206.4 | NP_077168.2 | 1 - 2 | 32 | 62 | |
GenBank mRNA | AK003354.1 | - | 1 - 2 | 33 | 62 | |
BC002128.1 | - | 1 - 2 | 35 | 3590767 | 62 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK003354.1,BC002128.1,NM_024206.4 transcripts.
This assay is in the 5'UTR region of the AK003354.1,BC002128.1,NM_024206.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
