Assay Details
Gene Symbol: Emid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080595.2 | NP_542162.1 | 4 - 5 | 391 | 61 | |
GenBank mRNA | AJ416093.1 | - | 4 - 5 | 391 | 61 | |
AK153702.1 | - | 4 - 5 | 410 | 61 | ||
BC130023.1 | - | 4 - 5 | 396 | 40140754 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
