Assay Details
Gene Symbol: Emid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC012248.1 | - | 5 - 6 | 308 | 56 | |
BC021849.1 | - | 4 - 5 | 210 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
