Assay Details
| Gene Symbol: Rpf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042556.1 | NP_001036021.1 | 7 - 8 | 911 | 109 | |
| NM_023323.3 | NP_075812.3 | 7 - 8 | 561 | 109 | ||
| GenBank mRNA | AB041810.1 | - | 7 - 8 | 941 | 109 | |
| AK010776.1 | - | 7 - 8 | 555 | 109 | ||
| AK011644.1 | - | 7 - 8 | 539 | 109 | ||
| AK012188.1 | - | 7 - 8 | 561 | 109 | ||
| AK012641.1 | - | 7 - 8 | 540 | 109 | ||
| AK013395.1 | - | 7 - 8 | 536 | 109 | ||
| AK017678.1 | - | 7 - 8 | 532 | 109 | ||
| AK020030.1 | - | 7 - 8 | 522 | 109 | ||
| AK051135.1 | - | 7 - 8 | 912 | 109 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpf2
Genomic Map