Assay Details
Gene Symbol: Tpd52l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK131855.1 | - | 4 - 5 | 474 | 103 | |
BC171924.1 | - | 4 - 5 | 464 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
