Assay Details
| Gene Symbol: Eldr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110421.1 | 3 - 4 | 358 | 106 | ||
| GenBank mRNA | BI990615.1 | - | 3 - 4 | 158 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eldr
Genomic Map