Assay Details
| Gene Symbol: Armc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034858.3 | NP_001030030.2 | 2 - 3 | 443 | 64 | |
| GenBank mRNA | BM942469.1 | - | 2 - 3 | 426 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Armc2
Genomic Map