Assay Details
| Gene Symbol: BC021785 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001489.3 | NP_001001489.1 | 3 - 4 | 549 | 60 | |
| GenBank mRNA | AK085266.1 | - | 3 - 4 | 550 | 60 | |
| BC094014.1 | - | 2 - 3 | 276 | 60 | ||
| BC147384.1 | - | 3 - 4 | 489 | 60 | ||
| BC147385.1 | - | 3 - 4 | 489 | 60 | ||
More Information
Set Membership: |
|

BC021785
Genomic Map