Assay Details
Gene Symbol: Fam184a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081428.1 | NP_001074897.1 | 15 - 16 | 3071 | 72 | |
GenBank mRNA | AK016375.1 | - | 6 - 7 | 800 | 72 | |
AK139673.1 | - | 10 - 11 | 1562 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
