Assay Details
| Gene Symbol: Lin28b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK133928.1 | - | 1 - 2 | 174 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK133928.1 transcripts.
Set Membership: |
|

Lin28b
Genomic Map