Assay Details
Gene Symbol: B230208H11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038027.1 | 2 - 3 | 644 | 95 | ||
GenBank mRNA | AK045519.1 | - | 2 - 3 | 608 | 95 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | utrophin | Utrn |
AA589569, DRP, Dmdl |
-51.9kb | Sense | Chr. 10 - 12382187 - 12869685 on NCBI Build GRCm38 |
Downstream | syntaxin 11 | Stx11 | 5830405C08Rik | 17.5kb | Sense | Chr. 10 - 12939983 - 12964500 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 754311 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 282554 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 9680 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 55074 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105427 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P105427 | - | - |
Whole Mouse Genome G4122A | A_51_P151516 | NR_038027.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P151516 | NR_038028.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P151516 | AK045519.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P151516 | AK045934.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128936 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103954 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P105427 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08308244 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08257813 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08487845 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948143 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08728599 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948262 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948252 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948193 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08324928 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948148 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948150 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08486638 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948240 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948151 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948166 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948146 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948248 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08625866 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08934363 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948214 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08244679 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948266 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08275661 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948210 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948215 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08244678 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948247 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948196 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948253 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08687255 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08275664 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948245 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948269 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948260 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948257 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08275660 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948221 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948212 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948139 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08731839 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948220 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948273 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948227 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948216 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08919395 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08630203 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08934359 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948141 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948140 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948176 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08934358 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948268 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08934362 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08599647 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948149 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948276 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08257817 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948203 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08315073 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08630199 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08791123 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948175 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948178 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948243 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948225 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948264 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08727156 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948204 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08580884 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948205 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26948170 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Clariom S Array | PSR1000012825.mm.1 | NR_038027.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000012825.mm.1 | NR_038027.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000012825.mm.1 | NR_038028.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000012825.mm.1 | NR_038028.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000012825.mm.1 | TC1000001760.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000012826.mm.1 | NR_038027.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000012826.mm.1 | NR_038027.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000012826.mm.1 | NR_038028.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000012826.mm.1 | NR_038028.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000012826.mm.1 | TC1000001760.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000012827.mm.1 | NR_038027.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000012827.mm.1 | NR_038027.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000012827.mm.1 | NR_038028.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000012827.mm.1 | NR_038028.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000012827.mm.1 | TC1000001760.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000012828.mm.1 | NR_038027.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000012828.mm.1 | NR_038027.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000012828.mm.1 | TC1000001760.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 337933 | - | - |
