Assay Details
Gene Symbol: D630024D03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_102312.1 | 4 - 5 | 613 | 75 | ||
NR_102314.1 | 3 - 4 | 461 | 75 | |||
GenBank mRNA | AK085420.1 | - | 4 - 5 | 591 | 75 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2334424 | AK158188.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 15286 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 953805 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 173140 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 44823 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 79206 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 40198 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 50784 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109596 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124545 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685159 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685079 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685167 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685118 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09524084 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27684897 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09017531 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27684918 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685150 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685062 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685114 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09290046 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685097 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685144 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685149 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685013 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685103 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27684920 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685164 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685070 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685140 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685156 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685063 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685092 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27684905 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685137 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09443546 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685141 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08993262 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09638555 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09627927 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09151181 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685008 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685077 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685112 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685117 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685075 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27684917 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27684906 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09267772 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685088 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685090 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685158 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09103215 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09443542 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685146 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685082 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09524067 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685110 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27684994 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685060 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685065 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09151172 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27684992 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27684930 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685129 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685120 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27684997 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685138 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685052 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09315461 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685002 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685131 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09103219 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27684999 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27684991 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685152 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685094 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09164927 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685124 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27684916 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685084 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685012 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685160 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09315459 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685125 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685102 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685053 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27684900 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27684901 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27684926 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09113994 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09689714 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09315465 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685115 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27684924 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685122 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27684914 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685087 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09570920 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09113991 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09518538 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27684899 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685057 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685011 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27684995 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685148 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09518530 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685058 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27685106 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1444315_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1444315_at | - | - |
Mouse Genome 430 2.0 Array | 1444315_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100022929.mm.1 | NR_102312.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100022929.mm.1 | NR_102314.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100022929.mm.1 | TC1100002464.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100022930.mm.1 | NR_102312.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100022930.mm.1 | NR_102314.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100022930.mm.1 | TC1100002464.mm.1 | MTA TC |
Mouse Expression Set 430 | 1444315_at | - | - |
Mouse Expression Set 430 | 1444315_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1444315_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1444315_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 320778 | - | - |
