Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400028619.mm.1 |
TC0400003525.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400028619.mm.1 |
NR_015560.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400028619.mm.1 |
NR_015560.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400028620.mm.1 |
TC0400003525.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400028620.mm.1 |
NR_015560.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400028620.mm.1 |
NR_015560.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400028612.mm.1 |
TC0400003525.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400028612.mm.1 |
NR_015560.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400028612.mm.1 |
NR_015560.1 |
Refseq |