Assay Details
| Gene Symbol: Ttll3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133923.6 | NP_598684.4 | 12 - 13 | 2404 | 67 | |
| GenBank mRNA | AK080321.1 | - | 8 - 9 | 1818 | 67 | |
| AM690746.1 | - | 12 - 13 | 2404 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ttll3
Genomic Map