Assay Details
| Gene Symbol: Gm10069 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028592.1 | 1 - 2 | 625 | 135 | ||
| NR_028593.1 | 1 - 2 | 625 | 135 | |||
| GenBank mRNA | AK029950.1 | - | 1 - 2 | 327 | 135 | |
| AK046088.1 | - | 1 - 2 | 581 | 135 | ||
| AK086937.1 | - | 1 - 2 | 626 | 135 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK046088.1, AK086937.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P110818 | AK029950.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P110818 | NR_028592.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 225892 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 337916 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 212771 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 412682 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 972841 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 87559 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P128595 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P159772 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P162499 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P110818 | AK029950.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P110818 | NR_028592.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06065913 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06065832 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01444497 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01444507 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P159772 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P162499 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P128595 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110818 | AK029950.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110818 | NR_028592.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P128595 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P162499 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P159772 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24839125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24839121 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05117004 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24839120 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05795283 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05117002 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05738736 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05702507 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05638234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05425245 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05653756 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24839115 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24839126 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10542106 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10542104 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10515884 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10521690 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10447702 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600012330.mm.1 | TC0600001510.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600012330.mm.1 | NR_028592.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600012330.mm.1 | NR_028592.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600012330.mm.1 | NR_028593.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600012330.mm.1 | NR_028593.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600012332.mm.1 | TC0600001510.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600012332.mm.1 | NR_028592.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600012332.mm.1 | NR_028592.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600012332.mm.1 | NR_028593.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600012332.mm.1 | NR_028593.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600012333.mm.1 | TC0600001510.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600012333.mm.1 | NR_028592.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600012333.mm.1 | NR_028592.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600012333.mm.1 | NR_028593.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600012333.mm.1 | NR_028593.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600012334.mm.1 | TC0600001510.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600012334.mm.1 | NR_028592.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600012334.mm.1 | NR_028592.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600012334.mm.1 | NR_028593.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600012334.mm.1 | NR_028593.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600012329.mm.1 | TC0600001510.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600012329.mm.1 | NR_028592.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600012329.mm.1 | NR_028592.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600012329.mm.1 | NR_028593.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600012329.mm.1 | NR_028593.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 104780161 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 100780706 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 104780161 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 100780706 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 104780161 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100780706 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104780161 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1240960 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1247293 | - | - |

Gm10069
Genomic Map