Assay Details
Gene Symbol: Gm20125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038020.1 | 6 - 7 | 997 | 90 | ||
GenBank mRNA | AK029503.1 | - | 6 - 7 | 999 | 90 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA C330004P14 gene | C330004P14Rik | -1.1mb | Anti-sense | Chr. 10 - 15791777 - 15807065 on NCBI Build GRCm38 | |
Downstream |
Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 2 |
Cited2 |
AI835299, ER154-like, Mrg1, Msg2, p35srj |
763.7kb | Anti-sense | Chr. 10 - 17723205 - 17725674 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
