Assay Details
Gene Symbol: Gm20125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038020.1 | 7 - 8 | 1152 | 130 | ||
GenBank mRNA | AK029503.1 | - | 7 - 8 | 1154 | 130 | |
AK136184.1 | - | 1 - 2 | 122 | 130 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA C330004P14 gene | C330004P14Rik | -1.1mb | Anti-sense | Chr. 10 - 15791777 - 15807065 on NCBI Build GRCm38 | |
Downstream |
Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 2 |
Cited2 |
AI835299, ER154-like, Mrg1, Msg2, p35srj |
767.2kb | Anti-sense | Chr. 10 - 17723205 - 17725674 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
