Assay Details
| Gene Symbol: Myo1g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178440.4 | NP_848534.2 | 19 - 20 | 2753 | 68 | |
| GenBank mRNA | AF426468.1 | - | 1 - 2 | 45 | 68 | |
| AK030825.1 | - | 19 - 20 | 2754 | 68 | ||
| AK036461.1 | - | 19 - 20 | 2738 | 68 | ||
| AK042033.1 | - | 19 - 20 | 2706 | 68 | ||
| AK088011.1 | - | 19 - 20 | 2754 | 68 | ||
| AK152797.1 | - | 19 - 20 | 2747 | 68 | ||
| AK159700.1 | - | 19 - 20 | 2729 | 68 | ||
| AK171284.1 | - | 19 - 20 | 2747 | 68 | ||
| AK171505.1 | - | 19 - 20 | 2727 | 68 | ||
| BC028661.1 | - | 7 - 8 | 1041 | 68 | ||
| BC113755.1 | - | 19 - 20 | 2697 | 40061202 | 68 | |
| BC113954.1 | - | 19 - 20 | 2697 | 40061200 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Myo1g
Genomic Map