Assay Details
| Gene Symbol: Nsun4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028142.4 | NP_082418.1 | 4 - 5 | 848 | 85 | |
| GenBank mRNA | AK009285.1 | - | 3 - 4 | 558 | 85 | |
| AK012994.1 | - | 4 - 5 | 849 | 85 | ||
| BC024628.1 | - | 4 - 5 | 768 | 4165638 | 85 | |
| FJ882982.1 | - | 4 - 5 | 3394 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nsun4
Genomic Map