| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT MG-430 PM Array Plate |
1457668_PM_x_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10507557 |
- |
- |
| Mouse Genome 430 2.0 Array |
1457668_PM_x_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1439898_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1439898_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1457668_x_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
100041290_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
100041290_PM_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0400009978.mm.1 |
NR_029469.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400009978.mm.1 |
NR_029469.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400009978.mm.1 |
TC0400001312.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400009973.mm.1 |
NM_026932.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400009973.mm.1 |
NM_026932.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400009973.mm.1 |
NR_029469.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400009973.mm.1 |
NR_029469.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400009973.mm.1 |
TC0400001312.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400009977.mm.1 |
NR_029469.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400009977.mm.1 |
NR_029469.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400009977.mm.1 |
TC0400001312.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1457668_x_at |
- |
- |
| Mouse Expression Set 430 |
1439898_PM_at |
- |
- |
| Mouse Expression Set 430 |
1457668_PM_x_at |
- |
- |
| Mouse Expression Set 430 |
1439898_at |
- |
- |
| Murine Genome U74v2 Set |
133836_PM_at |
- |
- |
| Murine Genome U74v2 Set |
133836_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1457668_PM_x_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1457668_x_at |
- |
- |