Assay Details
Gene Symbol: Esr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK036627.1 | - | 1 - 2 | 110 | 89 | |
EU791538.1 | - | 1 - 2 | 63 | 89 | ||
EU791540.1 | - | 1 - 2 | 188 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK036627.1, EU791538.1, EU791540.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
