Assay Details
| Gene Symbol: Esr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AI386103.1 | - | 1 - 2 | 151 | 74 | |
| AK041525.1 | - | 1 - 2 | 149 | 74 | ||
| AK139745.1 | - | 1 - 2 | 561 | 74 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK041525.1, AK139745.1 transcripts.
Set Membership: |
|

Esr1
Genomic Map