Assay Details
| Gene Symbol: Fgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010208.4 | NP_034338.3 | 2 - 3 | 234 | 72 | |
| GenBank mRNA | AK036438.1 | - | 2 - 3 | 156 | 72 | |
| AK036476.1 | - | 2 - 3 | 406 | 72 | ||
| AK038141.1 | - | 2 - 3 | 140 | 72 | ||
| AK089812.1 | - | 2 - 3 | 204 | 72 | ||
| AK155902.1 | - | 2 - 3 | 210 | 72 | ||
| AK157640.1 | - | 188 | 72 | |||
| AK171824.1 | - | 2 - 3 | 188 | 72 | ||
| BC057863.1 | - | 2 - 3 | 103 | 5317449 | 72 | |
| X16440.1 | - | 2 - 3 | 169 | 72 | ||
| X52191.1 | - | 2 - 3 | 141 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK036438.1, AK036476.1, AK038141.1, AK089812.1, AK155902.1, AK171824.1, BC057863.1, NM_010208.4, X16440.1, X52191.1 transcripts.
Set Membership: |
|

Fgr
Genomic Map