Assay Details
Gene Symbol: Fgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010208.4 | NP_034338.3 | 8 - 9 | 1054 | 64 | |
GenBank mRNA | AK036438.1 | - | 8 - 9 | 976 | 64 | |
AK036476.1 | - | 8 - 9 | 1226 | 64 | ||
AK038141.1 | - | 8 - 9 | 960 | 64 | ||
AK089812.1 | - | 8 - 9 | 1024 | 64 | ||
AK155902.1 | - | 8 - 9 | 1030 | 64 | ||
AK157640.1 | - | 1008 | 64 | |||
AK171824.1 | - | 8 - 9 | 1008 | 64 | ||
BC057863.1 | - | 8 - 9 | 923 | 5317449 | 64 | |
X16440.1 | - | 8 - 9 | 989 | 64 | ||
X52191.1 | - | 8 - 9 | 961 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
