Assay Details
| Gene Symbol: Gm12977 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016328.1 | - | 1 - 2 | 200 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 30019 |
Gm30019 | -96.4kb | Anti-sense | Chr. 4 - 133768128 - 133775447 on NCBI Build GRCm38 | |
| Downstream | high mobility group nucleosomal binding domain 2 | Hmgn2 |
HMG-17, Hmg17 |
88.9kb | Anti-sense | Chr. 4 - 133964739 - 133967991 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm12977
Genomic Map