Assay Details
| Gene Symbol: Gm11946 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK035589.1 | - | 2 - 3 | 1113 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | smoothelin | Smtn | smsmo | Anti-sense | Chr. 11 - 3517522 - 3539654 on NCBI Build GRCm38 | |
| Downstream | taurine upregulated gene 1 | Tug1 | AI316828 | 103.6kb | Anti-sense | Chr. 11 - 3639785 - 3648814 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm11946
Genomic Map