Assay Details
| Gene Symbol: Pde7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013875.5 | NP_038903.3 | 8 - 9 | 1080 | 140 | |
| GenBank mRNA | AF190639.1 | - | 8 - 9 | 861 | 140 | |
| AJ251859.1 | - | 8 - 9 | 1132 | 140 | ||
| AK035385.1 | - | 8 - 9 | 1092 | 140 | ||
| AK049910.1 | - | 8 - 9 | 1136 | 140 | ||
| BC130267.1 | - | 8 - 9 | 961 | 40143060 | 140 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pde7b
Genomic Map