Assay Details
| Gene Symbol: Ewsr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283061.1 | NP_001269990.1 | 3 - 4 | 164 | 71 | |
| NM_001283062.1 | NP_001269991.1 | 3 - 4 | 164 | 71 | ||
| NM_001283063.1 | NP_001269992.1 | 3 - 4 | 164 | 71 | ||
| NM_007968.3 | NP_031994.2 | 3 - 4 | 164 | 71 | ||
| GenBank mRNA | AK019460.1 | - | 3 - 4 | 113 | 71 | |
| AK046434.1 | - | 3 - 4 | 173 | 71 | ||
| AK049743.1 | - | 3 - 4 | 144 | 71 | ||
| AK146791.1 | - | 3 - 4 | 140 | 71 | ||
| AK147103.1 | - | 3 - 4 | 131 | 71 | ||
| AK147909.1 | - | 3 - 4 | 121 | 71 | ||
| AK150345.1 | - | 3 - 4 | 122 | 71 | ||
| AK151625.1 | - | 3 - 4 | 135 | 71 | ||
| AK153276.1 | - | 3 - 4 | 122 | 71 | ||
| BC068226.1 | - | 3 - 4 | 131 | 5707017 | 71 | |
| X79233.1 | - | 3 - 4 | 153 | 71 | ||
More Information
Set Membership: |
|

Ewsr1
Genomic Map