Assay Details
| Gene Symbol: Polm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017401.2 | NP_059097.2 | 3 - 4 | 734 | 83 | |
| GenBank mRNA | AF176098.1 | - | 3 - 4 | 507 | 83 | |
| AJ251804.2 | - | 3 - 4 | 478 | 83 | ||
| BC056213.1 | - | 3 - 4 | 526 | 30024844 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Polm
Genomic Map