Assay Details
| Gene Symbol: Inpp5j | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172439.3 | NP_766027.2 | 5 - 6 | 1671 | 75 | |
| GenBank mRNA | AK034272.1 | - | 5 - 6 | 1672 | 75 | |
| AK136908.1 | - | 5 - 6 | 1658 | 75 | ||
| AK165718.1 | - | 4 - 5 | 2199 | 75 | ||
| AK170114.1 | - | 5 - 6 | 1637 | 75 | ||
| AK171029.1 | - | 5 - 6 | 1632 | 75 | ||
| BC022915.1 | - | 5 - 6 | 1624 | 75 | ||
| BC046561.1 | - | 4 - 5 | 484 | 75 | ||
| BC051452.1 | - | 5 - 6 | 1641 | 75 | ||
| BC131634.1 | - | 5 - 6 | 1666 | 40096940 | 75 | |
| BC131635.1 | - | 5 - 6 | 1663 | 40096943 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Inpp5j
Genomic Map