Assay Details
| Gene Symbol: Inpp5j | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172439.3 | NP_766027.2 | 8 - 9 | 2067 | 61 | |
| GenBank mRNA | AK034272.1 | - | 8 - 9 | 2068 | 61 | |
| AK136908.1 | - | 8 - 9 | 2054 | 61 | ||
| AK165718.1 | - | 7 - 8 | 2595 | 61 | ||
| AK170114.1 | - | 8 - 9 | 2033 | 61 | ||
| AK171029.1 | - | 8 - 9 | 2028 | 61 | ||
| BC022915.1 | - | 8 - 9 | 2020 | 61 | ||
| BC046561.1 | - | 7 - 8 | 880 | 61 | ||
| BC051452.1 | - | 8 - 9 | 2036 | 61 | ||
| BC131634.1 | - | 8 - 9 | 2062 | 40096940 | 61 | |
| BC131635.1 | - | 8 - 9 | 2059 | 40096943 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Inpp5j
Genomic Map