Assay Details
| Gene Symbol: Mecr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025297.2 | NP_079573.2 | 6 - 7 | 790 | 84 | |
| GenBank mRNA | BC003864.1 | - | 6 - 7 | 790 | 3488760 | 84 |
| BQ746166.1 | - | 3 - 4 | 231 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mecr
Genomic Map