Assay Details
| Gene Symbol: Mecr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025297.2 | NP_079573.2 | 8 - 9 | 919 | 68 | |
| GenBank mRNA | AK002317.1 | - | 8 - 9 | 920 | 68 | |
| AK002533.1 | - | 8 - 9 | 919 | 68 | ||
| BC003864.1 | - | 8 - 9 | 919 | 3488760 | 68 | |
| BQ746166.1 | - | 5 - 6 | 360 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mecr
Genomic Map