Assay Details
| Gene Symbol: Rps6ka1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285505.1 | NP_001272434.1 | 15 - 16 | 1471 | 63 | |
| NM_001285506.1 | NP_001272435.1 | 14 - 15 | 1355 | 63 | ||
| NM_009097.5 | NP_033123.2 | 15 - 16 | 1504 | 63 | ||
| GenBank mRNA | AK132856.1 | - | 13 - 14 | 1541 | 63 | |
| AK148208.1 | - | 15 - 16 | 1497 | 63 | ||
| AK167790.1 | - | 14 - 15 | 1342 | 63 | ||
| BC049076.1 | - | 15 - 16 | 1455 | 63 | ||
| BC094470.1 | - | 15 - 16 | 1457 | 5098650 | 63 | |
| M28489.1 | - | 15 - 16 | 1457 | 63 | ||
More Information
Set Membership: |
|

Rps6ka1
Genomic Map