Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400026566.mm.1 |
TC0400003331.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400026566.mm.1 |
NM_001122958.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026566.mm.1 |
NM_001122958.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026566.mm.1 |
NM_001122959.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026566.mm.1 |
NM_001122959.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026566.mm.1 |
NM_009015.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026566.mm.1 |
NM_009015.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026565.mm.1 |
TC0400003331.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400026565.mm.1 |
NM_001122958.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026565.mm.1 |
NM_001122958.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026565.mm.1 |
NM_001122959.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026565.mm.1 |
NM_001122959.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026565.mm.1 |
NM_009015.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026565.mm.1 |
NM_009015.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026563.mm.1 |
TC0400003331.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400026563.mm.1 |
NM_001122958.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026563.mm.1 |
NM_001122958.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026563.mm.1 |
NM_001122959.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026563.mm.1 |
NM_001122959.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026563.mm.1 |
NM_009015.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026563.mm.1 |
NM_009015.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026564.mm.1 |
TC0400003331.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400026564.mm.1 |
NM_001122958.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026564.mm.1 |
NM_001122958.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026564.mm.1 |
NM_001122959.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026564.mm.1 |
NM_001122959.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026564.mm.1 |
NM_009015.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026564.mm.1 |
NM_009015.3 |
Refseq |