Assay Details
| Gene Symbol: Faah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010173.4 | NP_034303.3 | 7 - 8 | 1194 | 105 | |
| GenBank mRNA | AK004985.1 | - | 7 - 8 | 1020 | 105 | |
| AK032079.1 | - | 7 - 8 | 968 | 105 | ||
| AK079598.1 | - | 7 - 8 | 1195 | 105 | ||
| AK080603.1 | - | 7 - 8 | 1026 | 105 | ||
| AK138481.1 | - | 7 - 8 | 814 | 105 | ||
| BC006863.1 | - | 7 - 8 | 973 | 3595191 | 105 | |
| BC052321.1 | - | 7 - 8 | 1236 | 6515634 | 105 | |
| U82536.1 | - | 7 - 8 | 955 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Faah
Genomic Map