Assay Details
Gene Symbol: Ddx56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026538.3 | NP_080814.1 | 10 - 11 | 1303 | 65 | |
GenBank mRNA | AK011136.1 | - | 10 - 11 | 1313 | 65 | |
AK076047.1 | - | 10 - 11 | 1321 | 65 | ||
AK076975.1 | - | 10 - 11 | 1303 | 65 | ||
AK077514.1 | - | 10 - 11 | 1316 | 65 | ||
AK077953.1 | - | 5 - 6 | 550 | 65 | ||
AK145628.1 | - | 10 - 11 | 1344 | 65 | ||
AK159793.1 | - | 10 - 11 | 1307 | 65 | ||
AK169654.1 | - | 10 - 11 | 1318 | 65 | ||
AK185833.1 | - | 2 - 3 | 122 | 65 | ||
BC018291.1 | - | 10 - 11 | 1297 | 4190167 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
