Assay Details
| Gene Symbol: Bves | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024285.2 | NP_077247.1 | 2 - 3 | 353 | 66 | |
| GenBank mRNA | AF124510.1 | - | 1 - 2 | 214 | 66 | |
| AF204174.1 | - | 2 - 3 | 351 | 66 | ||
| BC132018.1 | - | 1 - 2 | 243 | 40130295 | 66 | |
| BC132044.1 | - | 1 - 2 | 243 | 40130321 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bves
Genomic Map