Assay Details
| Gene Symbol: Bves | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024285.2 | NP_077247.1 | 4 - 5 | 675 | 98 | |
| GenBank mRNA | AF124510.1 | - | 3 - 4 | 536 | 98 | |
| AF204174.1 | - | 4 - 5 | 673 | 98 | ||
| BC132018.1 | - | 3 - 4 | 565 | 40130295 | 98 | |
| BC132044.1 | - | 3 - 4 | 565 | 40130321 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bves
Genomic Map