Assay Details
| Gene Symbol: Guca2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008191.2 | NP_032217.2 | 2 - 3 | 349 | 96 | |
| GenBank mRNA | AK002364.1 | - | 2 - 3 | 321 | 96 | |
| BC024373.1 | - | 2 - 3 | 316 | 4971289 | 96 | |
| CF423670.1 | - | 2 - 3 | 362 | 96 | ||
| U67800.1 | - | 2 - 3 | 305 | 96 | ||
| U90727.1 | - | 2 - 3 | 351 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1957739 | NM_008191.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1957739 | AK002364.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1957739 | AV002130.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1957739 | AV003693.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1957739 | U67800.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 890464 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102884 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1957739 | NM_008191.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1957739 | AK002364.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1957739 | AV002130.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1957739 | AV003693.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1957739 | U67800.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P183414 | NM_008191.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P183414 | AK002364.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P183414 | AV002130.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P183414 | AV003693.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P183414 | U67800.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23246712 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23246697 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23246685 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23246687 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23246715 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23246677 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23246689 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23246704 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23246694 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23246698 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23246709 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23246691 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23246703 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23246682 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23246696 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23246701 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23246688 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23246707 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23246714 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23246718 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23246681 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23246700 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23246716 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23246693 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23246680 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23246711 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23246678 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23246706 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23246684 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23246708 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P102884 | - | - |
| Whole Mouse Genome G4122A | A_51_P183414 | NM_008191.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P183414 | AK002364.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P183414 | AV002130.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P183414 | AV003693.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P183414 | U67800.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112562 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P102884 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04097246 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03977276 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04097242 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03977268 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03977270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23246696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23246687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23246708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23246691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23246673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23246701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23246677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23246703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23246669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23246684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23246694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23246674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23246715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23246675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23246698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23246693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23246711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23246667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23246671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23246689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23246712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23246682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23246707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23246685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23246688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23246680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23246714 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03612124 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23246712 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03612104 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23246685 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03977272 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23246703 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03612126 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04071503 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04097244 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23246689 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23246701 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23246715 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23246667 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03464474 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23246693 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04071504 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23246675 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03424069 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23246694 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03612122 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23246698 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03977266 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23246673 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03612113 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04097250 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03612108 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23246706 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03612111 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03612119 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03612118 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04071509 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10515936 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515938 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515937 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1460233_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10515935 | - | - |
| Mouse Genome 430 2.0 Array | 1460233_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1460233_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14916_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14916_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400027880.mm.1 | TC0400003433.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400027880.mm.1 | NM_008191.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400027880.mm.1 | NM_008191.2 | Refseq |
| Mouse Expression Set 430 | 1460233_PM_at | - | - |
| Mouse Expression Set 430 | 1460233_at | - | - |
| Murine Genome U74v2 Set | 95569_at | - | - |
| Murine Genome U74v2 Set | 95569_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460233_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460233_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460233_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460233_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 652310 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4200014 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4200014 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004200014 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2744846 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2744846 | - | - |
| mouse-6 v1.1 (Illumina) | scl23889.4.1_100-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4200014 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl23889.4.1_100-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4200014 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2744846 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2744846 | - | - |

Guca2b
Genomic Map