Assay Details
| Gene Symbol: Tceb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013736.4 | NP_038764.2 | 1 - 2 | 94 | 128 | |
| GenBank mRNA | AK036592.1 | - | 1 - 2 | 95 | 128 | |
| AK145994.1 | - | 1 - 2 | 95 | 128 | ||
| AK147088.1 | - | 1 - 2 | 89 | 128 | ||
| BC049885.1 | - | 1 - 2 | 90 | 5387774 | 128 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tceb3
Genomic Map