Assay Details
| Gene Symbol: Tceb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013736.4 | NP_038764.2 | 6 - 7 | 1713 | 87 | |
| GenBank mRNA | AB025015.1 | - | 6 - 7 | 1697 | 87 | |
| AK036592.1 | - | 6 - 7 | 1714 | 87 | ||
| AK145994.1 | - | 6 - 7 | 1714 | 87 | ||
| AK147088.1 | - | 6 - 7 | 1708 | 87 | ||
| BC049885.1 | - | 6 - 7 | 1709 | 5387774 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tceb3
Genomic Map