Assay Details
Gene Symbol: Tceb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013736.4 | NP_038764.2 | 9 - 10 | 2104 | 91 | |
GenBank mRNA | AB025015.1 | - | 9 - 10 | 2088 | 91 | |
AK036592.1 | - | 9 - 10 | 2105 | 91 | ||
AK145994.1 | - | 9 - 10 | 2105 | 91 | ||
AK147088.1 | - | 9 - 10 | 2099 | 91 | ||
BC049885.1 | - | 9 - 10 | 2100 | 5387774 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
