Assay Details
| Gene Symbol: Hmgcl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008254.2 | NP_032280.2 | 2 - 3 | 166 | 104 | |
| GenBank mRNA | AK077842.1 | - | 2 - 3 | 155 | 104 | |
| AK132035.1 | - | 2 - 3 | 155 | 104 | ||
| AK144768.1 | - | 2 - 3 | 104 | |||
| AK145251.1 | - | 2 - 3 | 104 | |||
| AK154033.1 | - | 2 - 3 | 155 | 104 | ||
| BB852743.1 | - | 2 - 3 | 104 | |||
| BC025440.1 | - | 2 - 3 | 153 | 5049759 | 104 | |
| S65036.1 | - | 2 - 3 | 154 | 104 | ||
More Information
Set Membership: |
|

Hmgcl
Genomic Map