Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400009019.mm.1 |
NR_002862.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400009019.mm.1 |
TC1400001136.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400010535.mm.1 |
NM_130881.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010535.mm.1 |
NM_130881.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010535.mm.1 |
NM_148917.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010535.mm.1 |
NM_148917.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010535.mm.1 |
TC0400001395.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400010533.mm.1 |
NM_130881.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010533.mm.1 |
NM_130881.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010533.mm.1 |
NM_148917.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010533.mm.1 |
NM_148917.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010533.mm.1 |
TC0400001395.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400010534.mm.1 |
NM_130881.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010534.mm.1 |
NM_130881.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010534.mm.1 |
NM_148917.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010534.mm.1 |
NM_148917.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010534.mm.1 |
TC0400001395.mm.1 |
MTA TC |