Assay Details
| Gene Symbol: Slc2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011400.3 | NP_035530.2 | 4 - 5 | 722 | 66 | |
| GenBank mRNA | AK089353.1 | - | 4 - 5 | 664 | 66 | |
| AK155252.1 | - | 4 - 5 | 664 | 66 | ||
| AK170425.1 | - | 4 - 5 | 664 | 66 | ||
| AK170873.1 | - | 4 - 5 | 664 | 66 | ||
| BC055340.1 | - | 4 - 5 | 691 | 6808315 | 66 | |
| M22998.1 | - | 4 - 5 | 626 | 66 | ||
| M23384.1 | - | 4 - 5 | 701 | 66 | ||
| X69697.1 | - | 1 - 2 | 62 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc2a1
Genomic Map