Assay Details
| Gene Symbol: Slc2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011400.3 | NP_035530.2 | 9 - 10 | 1493 | 68 | |
| GenBank mRNA | AK155252.1 | - | 9 - 10 | 1435 | 68 | |
| AK170425.1 | - | 9 - 10 | 1435 | 68 | ||
| AK170873.1 | - | 9 - 10 | 1435 | 68 | ||
| BC055340.1 | - | 9 - 10 | 1462 | 6808315 | 68 | |
| M22998.1 | - | 9 - 10 | 1397 | 68 | ||
| M23384.1 | - | 9 - 10 | 1472 | 68 | ||
| S77924.1 | - | 1 - 2 | 215 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc2a1
Genomic Map