| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0X00015459.mm.1 |
NM_001126488.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00015459.mm.1 |
NM_001126488.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00015459.mm.1 |
TC0X00002446.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00015457.mm.1 |
NM_001126488.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00015457.mm.1 |
NM_001126488.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00015457.mm.1 |
TC0X00002446.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00015458.mm.1 |
NM_001126488.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00015458.mm.1 |
NM_001126488.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00015458.mm.1 |
TC0X00002446.mm.1 |
MTA TC |